Axygen® AxyPrep™ Mag Plasmid Kit, Corning
Supplier: Corning
MAG-P-10
10159-702CS 318.55 USD
10159-702
Axygen® AxyPrep™ Mag Plasmid Kit, Corning
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The AxyPrep™ Mag Plasmid purification kit utilizes a magnetic bead technology for high-throughput purification of plasmid DNA from E.coli cells.
AxyPrep™ Mag Plasmid kit can also be used with fosmid and BAC vector-based constructs ranging from 5kb to 150kb.