SHOWA BO600 Ergonomic Design, Polyurethane Fingertip Coated Product Protection Gloves, White, Showa
Supplier: Showa
BO600-XL BO600-S BO600-M BO600-L
10159-640CS 372.93 USD
10159-640 10159-650 10159-652 10159-654
SHOWA BO600 Ergonomic Design, Polyurethane Fingertip Coated Product Protection Gloves, White, Showa
Gloves
Proprietary ergonomic design provides unequalled comfort and dexterity.
  • Polyurethane white fingertip coated
  • 13 gauge Nylon white liner
  • Ergonomic design
These gloves are used in clean environments. They are lightweight for delicate part handling and inspection, with a polyurethane fingertip coating.
Packaging: Individually poly-bagged.