VWR® Preprinted Flammable Warning Tape
Supplier: VWR International
VWR-T18-FLA-C
10159-644CS 14.48 USD
10159-644
VWR® Preprinted Flammable Warning Tape
Tapes
VWR® Preprinted Flammable Warning Tape is imprinted with red ink on white tape and can be used on virtually any surface where visual warning is required.
  • Durable
  • Self-adhesive design
  • No residue
This durable, self-adhesive tape can withstand temperatures from -23 to 121°C (-10 to 250°F). The adhesive backing will peel off easily and leaves no residue.