Omega II Cap Safety Helmet, ERB Safety
Supplier: ERB INDUSTRIES, INC.
19951
10159-642CS 158.67 USD
10159-642
Omega II Cap Safety Helmet, ERB Safety
Head Protection Safety Helmets
Omega II cap safety helmets with ratchet knob adjustment.
  • 6-point woven nylon ratchet suspension
  • Accomodates chin strap
  • Certified when headband is installed in reverse position
  • Moisture-wicking brow pad is removable, ashable, and replaceable
  • Goggle retaining slots on suspension
  • Fits head sizes 6.5 - 8
Brow pad is replaceable.
Accessory slots accommodate a wide variety of eye, face, and hearing protection.
Certifications: Meets ANSI/ISEA Z89.1, Type 1, Class C, E and G. SEI ceritified.
Accessories information: Accessories not inlcuded.