Anti-IgG Polyclonal Antibody (SureLight® P3)

Supplier: Columbia Biosciences
D7-110-1MG
10159-224EA 533.53 USD
10159-224
Anti-IgG Polyclonal Antibody (SureLight® P3)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: SureLight® P3
Epitope:
Host:
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR