Standard Methods Agar (Plate Count Agar) with Chloramphenicol, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
W43
10158-954PK 96.13 USD
10158-954
Standard Methods Agar (Plate Count Agar) with Chloramphenicol, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and enumeration of yeast and mold.
Standard Methods Agar with Chloramphenicol 100ug/L is recommended for the isolation and enumeration of yeast and mold.
Ordering information: Minimum order of 10 packs