Shaking Water Bath, OLS Aqua Pro, Grant
Supplier: GRANT USA INC.
OLS26US
10158-864EA 7595.9 USD
10158-864
Shaking Water Bath, OLS Aqua Pro, Grant
Baths Shaking Water Baths
Combined orbital and linear shaking mechanism of the OLS26 allows optimisation of aeration and shear forces mixing, for reproducible results.
  • Precision digital temperature control
  • Temperature range 0 ºC to 99 ºC operation
  • Two LED displays (individual displays and controls for temperature and shaking speed)
  • Easy changeover from linear to orbital shaking
  • Adjustable shaking speed and stroke length
  • Universal tray and polycarbonate lid included as standard
  • Drain tap for convenient emptying
The OLS26 offers orbital and linear motion in one bath. Simply rotate the tray carrier by 180° to change between the two shaking modes. No need to buy separate trays to achieve orbital or linear motion. OLS26 features a dual display showing temperature and shaking speed and has adjustable shaking speed/stroke length. 
Applications:
General use – defrosting, cooling/warming liquids, temperature control of samples
Life-science – microbiological assays, tissue studies, cell cultivation fermentation, bacterial culture, biochemical assays, enzyme assay
Industrial – materials testing, corrosion testing
Biopharm – solubility testing of medical coatings, dissolution, cooling crystallisation
Food & beverage – extractions, food digestion
Certifications: 120V models are CSA and CE approved.