Brute® Recycling Container, Rubbermaid® Commercial
Supplier: Janitorial Supplies
RCP263273BE
10158-642EA 127.69 USD
10158-642
Brute® Recycling Container, Rubbermaid® Commercial
Bins Waste Bins
Large-capacity receptacle makes great sense for both mobile-materials collection and centralized-bin stations.
  • Tested to 20000 cycles
  • Guaranteed to never fade, warp, crack or crush
  • Rust-, chip- and peel proof
  • Colors complement compliance, recycling and sortation needs
  • Contoured base handles improve grip and ergonomics, reducing strain and improving efficiency when emptying containers
  • Rim features integrated rib structure to increase strength and resist crushing
  • Stackable design enables nesting for easy transport and storage
  • Opening size (Ø): 22"
  • Open top
  • For indoor and outdoor use
Integrated cinches secure the liner, allowing for efficient knot free liner changes. All-plastic commercial-grade construction. Proprietary design constructed with the highest quality material plus a UV inhibitor ensures long life in even the most extreme commercial environments.
Rounded handles make lifting and moving easier, and are reinforced to resist tearing or damage from even the heaviest loads. Reinforced base is specifically engineered to be dragged over rough surfaces, extending life and overall durability even in the toughest environments.