VWR® 4-Place Mini Bead Mill Homogenizer
Supplier: VWR International
10158-558
10158-558EA 2547.49 USD
10158-558
VWR® 4-Place Mini Bead Mill Homogenizer
Homogenizers Benchtop Homogenizers
The VWR® 4-Place Mini Bead Mill Homogenizer is a compact and powerful bead mill homogenizer.
  • Specifically designed for grinding, lysing, and the homogenization of biological samples prior to molecular extraction
  • Ideal solution for releasing DNA, RNA, proteins, and enzymes from tough samples while still retaining molecular integrity
  • Offers a broad performance range
Using sample tubes pre-filled with a variety of lysing beads, the mill vigorously and uniformly shakes the tubes providing an efficient, consistent, high-yield and quality homogenization in less than 60 seconds. The VWR® 4-Place Mini Bead Mill supports simultaneous processing of samples in 4x0.5 ml, 4x1.5 ml, 4x2 ml, or 1x7 ml tubes. It does not require a cool down period between runs, allowing for non-stop use and high throughput. The unit features a convenient front loading design and safety lid closure.
Certifications: CE marked.
Ordering information: Includes the VWR® Mini Bead Mill Drive Unit, integrated 0.5 ml, 1.5 ml and 2 ml tube carriage, interchangeable 1x7 ml tube carriage, user manual, and two-year warranty. VWR® disposable bead tubes are sold separately.