VWR® Reinforced 2 ml Bead Mill Tubes
Supplier: VWR International
10158-556
10158-556PK 512.06 USD
10158-556
VWR® Reinforced 2 ml Bead Mill Tubes
Tubes Centrifuge Tubes
VWR® Reinforced 2 ml tubes are specially formulated for bead beating applications.
  • Polypropylene construction
  • Reinforced to withstand high forces
  • Shatter-resistant
Each 2 ml polypropylene screw cap tube features a skirted base and secure O-ring.
Bead beating tubes are designed to withstand the high forces generated through bead milling. They are certified RNase and DNase free to ensure high analyte integrity.
VWR® Reinforced 2 ml tubes are compatible with the VWR® 4-Place Mini Bead Mill, Omni International Bead Ruptor 12 and Bead Ruptor 24, Next Advance Bullet Blender, Bertin Technologies Precellys 24 and Precellys Evolution, MP Biomedicals FastPrep® 24 and FastPrep® 24 5G bead mill homogenizers.
Certifications: RNase and DNase free.