Anti-MAP2K4 Rabbit Polyclonal Antibody

Supplier: Proteintech
51142-1-AP-150UL
10156-170EA 462.47 USD
10156-170
Anti-MAP2K4 Rabbit Polyclonal Antibody
Antibodies
The MEK4 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human MEK4. This antibody recognizes human, mouse antigen. The MEK4 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

IHC 1 : 50

Type: Primary
Antigen: MAP2K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR