Anti-C21ORF58 Rabbit Polyclonal Antibody

Supplier: Proteintech
25024-1-AP-150UL
10156-112EA 462.47 USD
10156-112
Anti-C21ORF58 Rabbit Polyclonal Antibody
Antibodies
The C21orf58 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C21orf58. This antibody recognizes human antigen. The C21orf58 antibody has been validated for the following applications: ELISA, IHC analysis.

WB 1 : 1000

Type: Primary
Antigen: C21ORF58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR