Anti-FOXD4 Rabbit Polyclonal Antibody

Supplier: Proteintech
24835-1-AP-150UL
10156-102EA 462.47 USD
10156-102
Anti-FOXD4 Rabbit Polyclonal Antibody
Antibodies
The FOXD4 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human FOXD4. This antibody recognizes human, mouse, rat antigen. The FOXD4 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600 ; IHC 1 : 50

Type: Primary
Antigen: FOXD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR