Anti-SMN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20451-1-AP-150UL
10156-058EA 462.47 USD
10156-058
Anti-SMN1 Rabbit Polyclonal Antibody
Antibodies
The SMN antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SMN. This antibody recognizes human, mouse antigen. The SMN antibody has been validated for the following applications: ELISA, WB, IHC, IF, IP analysis.

WB 1 : 1000

Type: Primary
Antigen: SMN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR