Anti-BCKDHB Rabbit Polyclonal Antibody

Supplier: Proteintech
13685-1-AP-150UL
10156-008EA 462.47 USD
10156-008
Anti-BCKDHB Rabbit Polyclonal Antibody
Antibodies
The BCKDHB antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human BCKDHB. This antibody recognizes human,mouse,rat antigen. The BCKDHB antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 1000

Type: Primary
Antigen: BCKDHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR