Anti-IFIT2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12604-1-AP-150UL
10155-992EA 462.47 USD
10155-992
Anti-IFIT2 Rabbit Polyclonal Antibody
Antibodies
The IFIT2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human IFIT2. This antibody recognizes human, mouse antigen. The IFIT2 antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.

LL

Type: Primary
Antigen: IFIT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR