Anti-EPSTI1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11627-1-AP-150UL
10155-982EA 462.47 USD
10155-982
Anti-EPSTI1 Rabbit Polyclonal Antibody
Antibodies
The EPSTI1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human EPSTI1. This antibody recognizes human, mouse antigen. The EPSTI1 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 600 ; IHC 1 : 50

Type: Primary
Antigen: EPSTI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR