Anti-AGTRAP Rabbit Polyclonal Antibody

Supplier: Proteintech
11559-1-AP-150UL
10155-978EA 462.47 USD
10155-978
Anti-AGTRAP Rabbit Polyclonal Antibody
Antibodies
The AGTRAP antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human AGTRAP. This antibody recognizes human,mouse,rat antigen. The AGTRAP antibody has been validated for the following applications: ELISA, IHC analysis.

WB 1 : 300 ; IHC 1 : 50

Type: Primary
Antigen: AGTRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR