Anti-MAPK7 Rabbit Polyclonal Antibody

Supplier: Proteintech
11164-1-AP-150UL
10155-970EA 462.47 USD
10155-970
Anti-MAPK7 Rabbit Polyclonal Antibody
Antibodies
The MAPK7 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human MAPK7. This antibody recognizes human,mouse,rat antigen. The MAPK7 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1200 ; IHC 1 : 100

Type: Primary
Antigen: MAPK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR