Anti-GOT1 Mouse Monoclonal Antibody [clone: 5A12E10]

Supplier: Proteintech
60317-1-IG-150UL
10155-904EA 462.47 USD
10155-904
Anti-GOT1 Mouse Monoclonal Antibody [clone: 5A12E10]
Antibodies
The GOT1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human GOT1. This antibody recognizes human antigen. The GOT1 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000

Type: Primary
Antigen: GOT1
Clonality: Monoclonal
Clone: 5A12E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR