Anti-VCP Mouse Monoclonal Antibody [clone: 2A4B10]

Supplier: Proteintech
60316-1-IG-150UL
10155-902EA 462.47 USD
10155-902
Anti-VCP Mouse Monoclonal Antibody [clone: 2A4B10]
Antibodies
The VCP antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human VCP. This antibody recognizes human, mouse antigen. The VCP antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 1000

Type: Primary
Antigen: VCP
Clonality: Monoclonal
Clone: 2A4B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR