Anti-TBL1XR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
55312-1-AP-150UL
10155-858EA 462.47 USD
10155-858
Anti-TBL1XR1 Rabbit Polyclonal Antibody
Antibodies
The TBL1XR1 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human TBL1XR1. This antibody recognizes human, mouse antigen. The TBL1XR1 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600 ; IF 1 : 25

Type: Primary
Antigen: TBL1XR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR