Anti-RCOR3 Rabbit Polyclonal Antibody

Supplier: Proteintech
55173-1-AP-150UL
10155-852EA 462.47 USD
10155-852
Anti-RCOR3 Rabbit Polyclonal Antibody
Antibodies
The RCOR3 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human RCOR3. This antibody recognizes human antigen. The RCOR3 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600 ; IP 1 : 300

Type: Primary
Antigen: RCOR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR