Anti-SIK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
51045-1-AP-150UL
10155-814EA 462.47 USD
10155-814
Anti-SIK1 Rabbit Polyclonal Antibody
Antibodies
The SIK1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human SIK1. This antibody recognizes human, mouse antigen. The SIK1 antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.

WB 1 : 300

Type: Primary
Antigen: SIK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR