Anti-MECR Rabbit Polyclonal Antibody

Supplier: Proteintech
51027-2-AP-150UL
10155-796EA 462.47 USD
10155-796
Anti-MECR Rabbit Polyclonal Antibody
Antibodies
The Mecr antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of mouse Mecr. This antibody recognizes human, mouse, rat antigen. The Mecr antibody has been validated for the following applications: ELISA, WB, IHC analysis.

IP 1 : 1000

Type: Primary
Antigen: MECR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR