Anti-ZBTB25 Rabbit Polyclonal Antibody

Supplier: Proteintech
25631-1-AP-150UL
10155-738EA 462.47 USD
10155-738
Anti-ZBTB25 Rabbit Polyclonal Antibody
Antibodies
The ZBTB25 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ZBTB25. This antibody recognizes human, mouse antigen. The ZBTB25 antibody has been validated for the following applications: ELISA, WB, IF analysis.

WB 1 : 600

Type: Primary
Antigen: ZBTB25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR