Anti-CDKN1B Rabbit Polyclonal Antibody

Supplier: Proteintech
25614-1-AP-150UL
10155-724EA 462.47 USD
10155-724
Anti-CDKN1B Rabbit Polyclonal Antibody
Antibodies
The P27; KIP1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human P27; KIP1. This antibody recognizes human, mouse antigen. The P27; KIP1 antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; WB 1 : 600

Type: Primary
Antigen: CDKN1B
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR