Anti-C1orf156 Rabbit Polyclonal Antibody

Supplier: Proteintech
25553-1-AP-150UL
10155-688EA 462.47 USD
10155-688
Anti-C1orf156 Rabbit Polyclonal Antibody
Antibodies
The C1orf156 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C1orf156. This antibody recognizes human antigen. The C1orf156 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000 ; IF 1 : 25 ; IHC 1 : 50 ; IP 1 : 300

Type: Primary
Antigen: C1orf156
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR