Anti-C18orf45 Rabbit Polyclonal Antibody

Supplier: Proteintech
25499-1-AP-150UL
10155-658EA 462.47 USD
10155-658
Anti-C18orf45 Rabbit Polyclonal Antibody
Antibodies
The C18orf45 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C18orf45. This antibody recognizes human antigen. The C18orf45 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600

Type: Primary
Antigen: C18orf45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR