Anti-DNAJC9 Rabbit Polyclonal Antibody

Supplier: Proteintech
25444-1-AP-150UL
10155-632EA 462.47 USD
10155-632
Anti-DNAJC9 Rabbit Polyclonal Antibody
Antibodies
The DNAJC9 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human DNAJC9. This antibody recognizes human antigen. The DNAJC9 antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; WB 1 : 600

Type: Primary
Antigen: DNAJC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR