Anti-OTUD6B Rabbit Polyclonal Antibody

Supplier: Proteintech
25430-1-AP-150UL
10155-618EA 462.47 USD
10155-618
Anti-OTUD6B Rabbit Polyclonal Antibody
Antibodies
The OTUD6B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human OTUD6B. This antibody recognizes human antigen. The OTUD6B antibody has been validated for the following applications: ELISA, WB, IHC analysis.

IHC 1 : 50

Type: Primary
Antigen: OTUD6B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR