Anti-CHD3 Rabbit Polyclonal Antibody

Supplier: Proteintech
25335-1-AP-150UL
10155-562EA 462.47 USD
10155-562
Anti-CHD3 Rabbit Polyclonal Antibody
Antibodies
The CHD3 Antibody from Proteintech is a rabbit polyclonal Antibody to a peptide of human CHD3. This Antibody recognizes human Antigen. The CHD3 Antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 1000

Type: Primary
Antigen: CHD3
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR