Anti-IQSEC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
25321-1-AP-150UL
10155-558EA 462.47 USD
10155-558
Anti-IQSEC2 Rabbit Polyclonal Antibody
Antibodies
The IQSEC2 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human IQSEC2. This Antibody recognizes human Antigen. The IQSEC2 Antibody has been validated for the following applications: ELISA, IHC analysis.

WB 1 : 300

Type: Primary
Antigen: IQSEC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR