Anti-GUCY2F Rabbit Polyclonal Antibody

Supplier: Proteintech
25252-1-AP-150UL
10155-524EA 462.47 USD
10155-524
Anti-GUCY2F Rabbit Polyclonal Antibody
Antibodies
The GUCY2F Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human GUCY2F. This Antibody recognizes human, mouse Antigen. The GUCY2F Antibody has been validated for the following applications: ELISA, WB, IHC analysis.

LL

Type: Primary
Antigen: GUCY2F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR