Anti-OARD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
25249-1-AP-150UL
10155-520EA 462.47 USD
10155-520
Anti-OARD1 Rabbit Polyclonal Antibody
Antibodies
The C6orf130 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human C6orf130. This Antibody recognizes human Antigen. The C6orf130 Antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 600 ; IHC 1 : 50

Type: Primary
Antigen: OARD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR