Anti-SLC10A2 Rabbit Polyclonal Antibody

Supplier: Proteintech
25245-1-AP-150UL
10155-516EA 462.47 USD
10155-516
Anti-SLC10A2 Rabbit Polyclonal Antibody
Antibodies
The ASBT Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human ASBT. This Antibody recognizes human, mouse Antigen. The ASBT Antibody has been validated for the following applications: ELISA, IP analysis.

WB 1 : 1000

Type: Primary
Antigen: SLC10A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR