Anti-AKAP12 Rabbit Polyclonal Antibody

Supplier: Proteintech
25199-1-AP-150UL
10155-484EA 462.47 USD
10155-484
Anti-AKAP12 Rabbit Polyclonal Antibody
Antibodies
The AKAP12 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human AKAP12. This antibody recognizes human, rat antigen. The AKAP12 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 300

Type: Primary
Antigen: AKAP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR