Anti-HIVEP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
25168-1-AP-150UL
10155-476EA 462.47 USD
10155-476
Anti-HIVEP1 Rabbit Polyclonal Antibody
Antibodies
The HIVEP1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human HIVEP1. This antibody recognizes human antigen. The HIVEP1 antibody has been validated for the following applications: ELISA, IHC analysis.

WB 1 : 300 ; IHC 1 : 50 ; IF 1 : 50

Type: Primary
Antigen: HIVEP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR