Anti-FIBCD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
25125-1-AP-150UL
10155-446EA 462.47 USD
10155-446
Anti-FIBCD1 Rabbit Polyclonal Antibody
Antibodies
The FIBCD1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human FIBCD1. This antibody recognizes human antigen. The FIBCD1 antibody has been validated for the following applications: ELISA, IHC analysis.

WB 1 : 1000

Type: Primary
Antigen: FIBCD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR