Anti-AWAT2 Rabbit Polyclonal Antibody

Supplier: Proteintech
25119-1-AP-150UL
10155-440EA 462.47 USD
10155-440
Anti-AWAT2 Rabbit Polyclonal Antibody
Antibodies
The AWAT2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human AWAT2. This antibody recognizes human, mouse, rat antigen. The AWAT2 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600 ; IHC 1 : 50 ; IF 1 : 25

Type: Primary
Antigen: AWAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR