Anti-MYO16 Rabbit Polyclonal Antibody

Supplier: Proteintech
25104-1-AP-150UL
10155-430EA 462.47 USD
10155-430
Anti-MYO16 Rabbit Polyclonal Antibody
Antibodies
The MYO16 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human MYO16. This antibody recognizes human, rat antigen. The MYO16 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600

Type: Primary
Antigen: MYO16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR