Anti-KIAA1530 Rabbit Polyclonal Antibody

Supplier: Proteintech
25054-1-AP-150UL
10155-404EA 462.47 USD
10155-404
Anti-KIAA1530 Rabbit Polyclonal Antibody
Antibodies
The KIAA1530 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human KIAA1530. This antibody recognizes human antigen. The KIAA1530 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600 ; IHC 1 : 50

Type: Primary
Antigen: KIAA1530
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR