Anti-SCCPDH Rabbit Polyclonal Antibody

Supplier: Proteintech
24941-1-AP-150UL
10155-366EA 462.47 USD
10155-366
Anti-SCCPDH Rabbit Polyclonal Antibody
Antibodies
The SCCPDH antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SCCPDH. This antibody recognizes human, mouse antigen. The SCCPDH antibody has been validated for the following applications: ELISA, IP, WB analysis.

WB 1 : 300

Type: Primary
Antigen: SCCPDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR