Anti-TMEM213 Rabbit Polyclonal Antibody

Supplier: Proteintech
24927-1-AP-150UL
10155-358EA 462.47 USD
10155-358
Anti-TMEM213 Rabbit Polyclonal Antibody
Antibodies
The TMEM213 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human TMEM213. This antibody recognizes human antigen. The TMEM213 antibody has been validated for the following applications: ELISA, IHC analysis.

WB 1 : 600

Type: Primary
Antigen: TMEM213
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR