Anti-LRP5 Rabbit Polyclonal Antibody

Supplier: Proteintech
24899-1-AP-150UL
10155-344EA 462.47 USD
10155-344
Anti-LRP5 Rabbit Polyclonal Antibody
Antibodies
The LRP5 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human LRP5. This antibody recognizes human antigen. The LRP5 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 600 ; IHC 1 : 200

Type: Primary
Antigen: LRP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR