Anti-PIK3C2B Rabbit Polyclonal Antibody

Supplier: Proteintech
24788-1-AP-150UL
10155-312EA 462.47 USD
10155-312
Anti-PIK3C2B Rabbit Polyclonal Antibody
Antibodies
The PIK3C2B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PIK3C2B. This antibody recognizes human antigen. The PIK3C2B antibody has been validated for the following applications: ELISA, WB, IP analysis.

WB 1 : 700 ; IHC 1 : 50

Type: Primary
Antigen: PIK3C2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR