Anti-TMEM35 Rabbit Polyclonal Antibody

Supplier: Proteintech
24786-1-AP-150UL
10155-310EA 462.47 USD
10155-310
Anti-TMEM35 Rabbit Polyclonal Antibody
Antibodies
The TMEM35 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human TMEM35. This antibody recognizes human, mouse antigen. The TMEM35 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600 ; IHC 1 : 50

Type: Primary
Antigen: TMEM35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR