Anti-CASP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24777-1-AP-150UL
10155-306EA 462.47 USD
10155-306
Anti-CASP2 Rabbit Polyclonal Antibody
Antibodies
The Caspase 2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human Caspase 2. This antibody recognizes human, mouse antigen. The Caspase 2 antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50

Type: Primary
Antigen: CASP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR