Anti-ODF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24736-1-AP-150UL
10155-294EA 462.47 USD
10155-294
Anti-ODF1 Rabbit Polyclonal Antibody
Antibodies
The ODF1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ODF1. This antibody recognizes human, rat, mouse antigen. The ODF1 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

IHC 1 : 50

Type: Primary
Antigen: ODF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR