Anti-C6orf70 Rabbit Polyclonal Antibody

Supplier: Proteintech
24690-1-AP-150UL
10155-278EA 462.47 USD
10155-278
Anti-C6orf70 Rabbit Polyclonal Antibody
Antibodies
The C6orf70 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C6orf70. This antibody recognizes human antigen. The C6orf70 antibody has been validated for the following applications: ELISA, IHC analysis.

WB 1 : 300 ; IHC 1 : 50 ; IF 1 : 25

Type: Primary
Antigen: C6orf70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR